Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse ABO, Blood Group A1B antigen, Clone: HE-24, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB11058716

Catalog No. NB11058716

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ABO, Blood Group A1B antigen Monoclonal antibody specifically detects ABO, Blood Group A1B antigen in Human samples. It is validated for Aggregation, Agglutination.
Specifications

Specifications

ABO, Blood Group A1B antigen
HE-24
Aggregation
Supernatant
ABO
Glycoprotein fraction isolated from saliva of secretors with blood group A.
Tissue culture supernatant
RUO
Primary
28
Human
Aggregation
Unconjugated
The hybridoma culture supernatant is 4 x concentrated by ultrafiltration using 100 kDa - cut off membrane. with No Preservative
A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT
Mouse
IgM
1 ml
Store at 4C. Do not freeze.
Monoclonal
The antibody HE-24 distinguishes blood group A1 B from A2 B. The specifity of the antibody HE-24 was verified on >1000 samples of blood.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only