Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-ACE-2, Clone: 881CT16.4.4, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232291
Catalog No. NBP232291
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ACE-2
881CT16.4.4
Western Blot 1:2000
Q9BYF1, Q9BYF1
ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Mouse
IgM
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot
Unconjugated
Crude ascites with 0.09% Sodium Azide
Ascites
ACE2
Purified His-tagged ACE2 protein was used to produced this monoclonal antibody.
92.463 kDa
100 ul
Immunology, Virology Bacteria and Parasites
Primary
59272