Learn More
anti-Acetylcholinesterase/ACHE, Clone: 684CT8.3.4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232061
Catalog No. NBP232061
Add to cart
Description
Description
Acetylcholinesterase/ACHE Monoclonal antibody specifically detects Acetylcholinesterase/ACHE in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
Acetylcholinesterase/ACHE
684CT8.3.4
Unconjugated
PBS with 0.09% Sodium Azide
Purified
ACHE
This Acetylcholinesterase antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 587-611 amino acids from the C-terminal region of human Acetylcholinesterase.
67.796 kDa
400 ul
Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
43
Western Blot
0.5 mg/mL
Western Blot 1:2000
P22303, Q29499
acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat