Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acrosin Antibody (ACR-2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500419UV
Description
Acrosin Monoclonal specifically detects Acrosin in Porcine, Bovine (Negative), Canine (Negative), Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Acrosin | |
Monoclonal | |
DyLight 350 | |
acrosin, ACRS, preproacrosin, proacrosin | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
The antibody ACR-2 reacts with various forms of porcine acrosin (55, 53, 45 and 35 kDa), a typical serine proteinase with trypsin-like specificity. Acrosin is stored in the acrosome of undamaged spermatozoa. | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
ACR-2 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
ACR | |
Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization. | |
0.1 mL | |
Signal Transduction | |
49 | |
Porcine, Bovine (Negative), Canine (Negative), Human (Negative) | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction