Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ACTH, Clone: 6Y8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP194713

Catalog No. NBP194713


Add to cart

Description

Description

ACTH Monoclonal antibody specifically detects ACTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ACTH
6Y8
Unconjugated
PBS and 30% Glycerol with 0.05% Sodium Azide
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein
Mouse
IgG1
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for the 1-24 amino acid sequence in ACTH and has no cross reaction with ACTH 17-39. The antibody has an affinity constant greater than 1011mol-1
Immunohistochemistry, Immunohistochemistry (Paraffin)
2 mg/mL
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1-4 ug/ml
Purified
POMC
Adrenocorticotropic hormone (ACTH) N-Terminal
Immunogen affinity purified
RUO
Primary
5443
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only