Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse Actin, Clone: 647, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142279

Catalog No. NBP142279


Add to cart

Description

Description

Actin Monoclonal antibody specifically detects Actin in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

Actin
647
Western Blot, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Supernatant
ACTA1
The antibody reacts with the Actin antigen.
Tissue culture supernatant
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
In addition to the intermediate filaments, the cytoskeleton contains contractile micro filaments. These proteins occur in most cell types, but more abundantly in muscle cells and myoepithelial cells. There are three types of contractile proteins: actin, myosin and tubuli. This antibody reacts in immunoblots with one single protein band which is corre?sponding to actin. Antigen origin: cell preparation of chicken skeletal muscle. Antigen location: cytoplasm.
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Unconjugated
Tissue culture supernatant with 0.1% Sodium Azide
ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3
Mouse
IgM
0.2 ml
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary
58
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only