Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Mouse AFAP, DyLight 350, Clone: 16F50, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222004UV
Description
AFAP Monoclonal specifically detects AFAP in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
AFAP | |
Monoclonal | |
DyLight 350 | |
Q12884 | |
AFAP1 | |
Human recombinant AFAP | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA | |
16F50 | |
Flow Cytometry, ELISA | |
actin filament associated protein 1, FLJ56849 | |
Mouse | |
Protein G purified | |
RUO | |
60312 | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only