Learn More
anti-AIM2, Clone: 812CT9.2.2.1, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232293
Catalog No. NBP232293
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
AIM2
812CT9.2.2.1
Western Blot 1:2000
O14862
absent in melanoma 2PYHIN4, interferon-inducible protein AIM2
Mouse
IgM
Unpurified
RUO
Primary
9447
Western Blot
Unconjugated
Crude ascites with 0.09% Sodium Azide
Ascites
AIM2
This AIM2 Antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-32 amino acids from human AIM2.
38.954 kDa
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human