Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-AK3, Clone: SJB3-36, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104261
Description
AK3 Monoclonal specifically detects AK3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
AK3 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10-1:500 | |
Adenylate kinase 3 alpha-like 1, adenylate kinase 3 like 1, adenylate kinase 3adenylate kinase 6, AK 3, AK6, AKL3L, AKL3L1, EC 2.7.4, EC 2.7.4.10, FIX, GTP:AMP phosphotransferase, mitochondrial | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human AK3 mAb, clone SJB3-36, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human AK3 protein. Clone SJB3-36 is specific against AK3. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
SJB3-36 | |
Unconjugated | |
NP_057366 | |
AK3 | |
Recombinant human AK3 (adenylaste kinase isozyme 3) purified from E. coli | |
0.1 mL | |
Signal Transduction | |
50808 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only