Learn More
anti-Akt1, Biotin, Clone: SB2H12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128593
Catalog No. NBP128593
Add to cart
Description
Description
AKT1 Monoclonal antibody specifically detects AKT1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
AKT1
SB2H12
Biotin
PBS with 0.05% Sodium Azide
AKT, EC 2.7.11, EC 2.7.11.1, PKBMGC99656, PRKBA, Protein kinase B, Proto-oncogene c-Akt, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, v-akt murine thymoma viral oncogene homolog 1
Mouse
IgG2b
0.05 mg
Store at 4C. Do not freeze.
Monoclonal
Human Akt/PKB
ELISA, Western Blot
0.5mg/mL
Western Blot 1:100-1:2000, ELISA 1:3000-1:5000
Purified
AKT1
The immunogen is a peptide made to the amino acid region AKT1
Protein A or G purified
RUO
Primary
207
Human