Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM82A Antibody, Alexa Fluor™ 750, Novus Biologicals™
Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP241121AF750
Description
FAM82A Polyclonal antibody specifically detects FAM82A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
FAM82A | |
Polyclonal | |
50mM Sodium Borate | |
Chicken | |
Peptide affinity purified | |
RUO | |
151393 | |
Store at 4°C in the dark. | |
IgY |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 750 | |
family with sequence similarity 82, member A1, member A, UNQ9371/PRO34163 | |
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human FAM82A1. The immunogen is located within amino acids 140 - 190 of FAM82A1. Amino Acid Squence: ITANTDTEEQSFPVPK | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction