Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Albumin HRP Polyclonal Antibody, Invitrogen™

Mouse Albumin Antibody, HRP conjugate, PA1-30126, from Invitrogen™. Species Reactivity: Mouse; Applications: ELISA, Western Blot.

Manufacturer:  Invitrogen PA130126

Catalog No. PA130126

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Mouse Albumin Antibody, HRP conjugate, PA1-30126, from Invitrogen. Species Reactivity: Mouse; Applications: ELISA, Western Blot.


Host Species: Goat
Cross Reactivity: Mouse
Isotype: IgG
Immunogen: Goats were immunized with mouse albumin. After multiple immunizations in Freund's adjuvant, the goats were bled and serum collected. The antibodies were immunoaffinity purified and conjugated with horseradish peroxidase (HRP).
Formulation: 1mg purified antibody (1mg/mL) in HEPES buffered saline with 0.2% BSA and 0.01% thimerosal; pH 7.0
Synonyms: Alb-1, Alb1
Uniprot Accession: P07724
Entrez GeneID: 11657
Gene Symbol: ALB
PA1-30126 detects Mouse Albumin from mouse samples. PA1-30126 has been successfully used in Western blot & ELISA applications. The immunogen for PA1-30126 was created by immunizing goats with mouse albumin. After multiple immunizations in Freunds adjuvant, the goats were bled and serum collected. The antibodies were immunoaffinity purified and conjugated with horseradish peroxidase (HRP).
Specifications

Specifications

Mouse Albumin
Polyclonal
1mg purified antibody (1mg/mL) in HEPES buffered saline with 0.2% BSA and 0.01% thimerosal; pH 7.0
Purified
ALB
Goats were immunized with mouse albumin. After multiple immunizations in Freund's adjuvant, the goats were bled and serum collected. The antibodies were immunoaffinity purified and conjugated with horseradish peroxidase (HRP).
1mg
Store at 4°C
Primary
11657
ELISA, Western Blot
HRP
P07724
Alb-1, Alb1
Goat
IgG
RUO
Primary
Polyclonal
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit