Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ALCAM/CD166, Clone: 3F8B12, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128894

Catalog No. NBP128894

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ALCAM/CD166
3F8B12
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
ALCAM
Purified recombinant fragment of ALCAM(aa405-524) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for ALCAM.
Western Blot, ELISA
Unconjugated
Ascites with 0.03% Sodium Azide
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Mouse
IgG2a
0.1 ml
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
214
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only