Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-alpha-Actinin 1, Clone: CB11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142554

Catalog No. NBP142554


Add to cart

Description

Description

alpha-Actinin 1 Monoclonal antibody specifically detects alpha-Actinin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

alpha-Actinin 1
CB11
Unconjugated
PBS solution containing 1.0% w/v BSA with 0.1% Sodium Azide
actinin 1 smooth muscle, actinin, alpha 1, alpha-actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1
Mouse
IgG1
0.05 mg
Angiogenesis, Cancer, Cytoskeleton Markers
Primary
87
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
0.1 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
ACTN1
The antibody reacts with the alpha-Actinin antigen.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The antibody is specific to the 100 kD a-actinin polypeptide and reacts with human and chicken cells. Monoclonal antibody to a-actinin is derived from the hybridoma produced by fusion between myeloma cells and Balb/c spleen cells. Purified a-actinin from chicken gizzard was used as an immunogen.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only