Learn More
anti-alpha-Fetoprotein/AFP, DyLight 488, Clone: 5E1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1102533G
Catalog No. NB1102533G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
alpha-Fetoprotein/AFP
50
Immunocytochemistry/Immunofluorescence
Purified
AFP
Full length native protein (purified) (Human).
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Reacts with different determinants of human alpha-fetoprotein receptor. Blocking alpha-fetoprotein and its receptor.
Immunocytochemistry, Immunofluorescence
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Mouse
IgG2a
0.2 ml
Cancer, GPCR, Stem Cell Markers
Primary
174
Human