Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Fodrin Antibody (AA6), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP142321
Description
Alpha Fodrin Monoclonal specifically detects Alpha Fodrin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Alpha Fodrin | |
Monoclonal | |
0.1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
SPTAN1 | |
The antibody reacts with the alpha-Fodrin antigen. | |
0.05 mg | |
Primary | |
The antibody is specific to the 240 kD a-fodrin molecule of all mammalian nonerythroid cells and chicken a-spectrin. The antibody cross reacts with human, rat, chicken, rabbit and guinea pig. Monoclonal antibody to a-fodrin is derived from the hybridoma produced by fusion between myeloma cells and Balb/c spleen cells. Chicken red blood cell membranes purified by hypotonic lysis and mechanical enucleation were used as immunogen. | |
Store at 4C. Do not freeze. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
AA6 | |
Unconjugated | |
Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
Mouse | |
Protein A or G purified | |
RUO | |
6709 | |
Human, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction