Learn More
Alpha Fodrin Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: 3D7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192689G
Catalog No. NBP192689G
Add to cart
Description
Description
Alpha Fodrin Monoclonal antibody specifically detects Alpha Fodrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Alpha Fodrin
3D7
Western Blot, Immunocytochemistry/Immunofluorescence
Q13813
Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
SPTAN1
Recombinant construct (expressed in and purified from E. coli) containing the 13th, 14th and 15th spectrin repeats of hiuman human alpha Fodrin (amino acids 1398-1685) [UniProt# Q13813].
240 kDa
0.1 ml
Neuroscience
Primary
6709