Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Fodrin Mouse anti-Human, Mouse, Rat, FITC, Clone: 3D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192689F

Catalog No. NBP192689F


Add to cart

Description

Description

Alpha Fodrin Monoclonal antibody specifically detects Alpha Fodrin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Alpha Fodrin
3D7
FITC
PBS with 0.05% Sodium Azide
Purified
SPTAN1
Recombinant construct (expressed in and purified from E. coli) containing the 13th, 14th and 15th spectrin repeats of hiuman human alpha Fodrin (amino acids 1398-1685) [UniProt# Q13813].
240 kDa
0.1 ml
Neuroscience
Primary
6709
Immunocytochemistry, Immunofluorescence
1mg/mL
Immunocytochemistry/Immunofluorescence
Q13813
Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only