Learn More
anti-Aminopeptidase N/CD13, Clone: 22A5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128429
Catalog No. NBP128429
Add to cart
Description
Description
Aminopeptidase N/CD13 Monoclonal antibody specifically detects Aminopeptidase N/CD13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-reported.
Specifications
Specifications
Aminopeptidase N/CD13
22A5
Unconjugated
Borate buffered saline, pH 8.2 with No Preservative
alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, AP-M, AP-N, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPNhAPN
Mouse
IgG2a Kappa
0.1 mg
Angiogenesis
Primary
290
Human
CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
0.1 mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-reported
Purified
ANPEP
Osteoclasts from osteoclastomas
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human CD13