Learn More
anti-Aminopeptidase N/CD13, FITC, Clone: 22A5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128430
Catalog No. NBP128430
Add to cart
Description
Description
Aminopeptidase N/CD13 Monoclonal antibody specifically detects Aminopeptidase N/CD13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
Aminopeptidase N/CD13
22A5
Flow Cytometry 10 uL/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
ANPEP
Osteoclasts from osteoclastomas
Protein A or G purified
RUO
Primary
290
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
FITC
PBS with Sodium Azide
alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, AP-M, AP-N, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPNhAPN
Mouse
IgG2a Kappa
1 ml
Store at 4C. Do not freeze.
Monoclonal
Human CD13