Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Androgen R/NR3C4, Clone: 1A9D12, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP151554

Catalog No. NBP151554

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Androgen R/NR3C4
1A9D12
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
AR
Purified recombinant fragment of AR (aa689-919) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Mouse
IgG1
0.1 ml
Breast Cancer, Cancer, Cellular Markers
Primary
367
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only