Learn More
anti-ApoE4, DyLight 405, Clone: 4E4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP149529V
Catalog No. NBP149529V
Add to cart
Description
Description
ApoE4 Monoclonal antibody specifically detects ApoE4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
ApoE4
40000
Western Blot, ELISA
P02649
Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571
Mouse
IgG1 Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody is specific for ApoE4. It does not cross-react with the ApoE2 or ApoE3 isoforms.
ELISA, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
APOE
Synthetic peptide made to an internal region of human ApoE4 (within residues 100-150) [Uniprot: P02649]
34 kDa
0.1 ml
Alzheimers Research, Apoptosis, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary
348
Human