Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Apolipoprotein A-IV/ApoA4, Clone: 1D4C11, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128896

Catalog No. NBP128896

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Apolipoprotein A-IV/ApoA4
1D4C11
Western Blot 1:500 - 1:2000, ELISA 1:10000
P06727
apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, MGC142154, MGC142156
Mouse
IgG2a
Unpurified
RUO
Primary
337
Human
Western Blot, ELISA
Unconjugated
Ascites with 0.03% Sodium Azide
Ascites
APOA4
Purified recombinant fragment of APOA4(aa 21-396) expressed in E. Coli.
45 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for APOA4.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only