Learn More
ATF6 Mouse anti-Human, Mouse, Rat, Porcine, Rabbit, DyLight 405, Clone: 70B1413.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP140256V
Catalog No. NBP140256V
Add to cart
Description
Description
ATF6 Monoclonal specifically detects ATF6 in Human, Mouse, Rat, Porcine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), Knockout Validated.
Specifications
Specifications
ATF6
70B1413.1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), Knockout Validated
P18850
activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha
Mouse
IgG1 Kappa
0.1 ml
Lipid and Metabolism, Signal Transduction, Unfolded Protein Response
Primary
22926
Human, Mouse, Rat, Porcine, Rabbit
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
ATF6
This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This ATF6 antibody detects both the full length and the cleaved/active protein.