Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Aurora C, Clone: 2B7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147663

Catalog No. NBP147663


Add to cart

Description

Description

Aurora C Monoclonal antibody specifically detects Aurora C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Aurora C
OTI2B7
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
AIE2AIK3, ARK3Serine/threonine-protein kinase aurora-C, AurC, aurora kinase CAurora/IPL1/Eg2 protein 2, Aurora/IPL1-related kinase 3, aurora-C, Aurora-related kinase 3, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 13 (aurora/IPL1-like), serine/threonine-protein kinase 13, STK13ARK-3
Mouse
IgG1
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for Homo sapiens aurora kinase C (AURKC), transcript variant 1.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
0.74 mg/mL
Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50
Affinity Purified
AURKC
Full-length protein expressed in 293T cell transfected with human AURKC expression vector
35.6 kDa
0.1 ml
Cell Cycle and Replication
Primary
6795
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only