Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Baculovirus Envelope gp64 protein, Clone: AcV1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP20020850UG
Description
Baculovirus Envelope gp64 protein Monoclonal specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Flow Cytometry.Specifications
Baculovirus Envelope gp64 protein | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000 | |
Mouse | |
Affinity Purified | |
RUO | |
Virus | |
IgG2a |
Flow Cytometry | |
AcV1 | |
Unconjugated | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
The immunogen for this antibody is baculovirus envelope gp64 protein. | |
50 μg | |
Primary | |
Store at 4C. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only