Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-BBS4, Clone: 1292CT845.130.218, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232308
Catalog No. NBP232308
Description
Description
BBS4 Monoclonal antibody specifically detects BBS4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
BBS4
1292CT845.130.218
Unconjugated
PBS with 0.09% Sodium Azide
Purified
BBS4
This BBS4 antibody is generated from a mice immunized with a recombinant protein between 1-240 amino acids from human BBS4.
58.282 kDa
400 ul
Stem Cell Markers, Vision
Primary
585
Western Blot
0.5 mg/mL
Western Blot 1:1000
Q96RK4
Bardet-Biedl syndrome 4, Bardet-Biedl syndrome 4 protein
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat