Learn More
anti-beta 2-Microglobulin, Biotin, Clone: F21-21, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP128313
Catalog No. NBP128313
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
beta 2-Microglobulin
F21-21
Biotin
PBS with 0.05% Sodium Azide
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Mouse
IgG1 Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
The beta2-microglobulin subunit (Mr approx. 14 kDa) of avian MHC Class I molecules
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.5mg/mL
Western Blot 1:100-1:2000, Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunocytochemistry
Purified
B2M
Affinity purified chicken MHC-I molecules
Protein A or G purified
RUO
Primary
567
Avian, Chicken, Turkey