Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BMI-1, FITC, Clone: LLBmi1-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196140F

Catalog No. NBP196140F


Add to cart

Description

Description

BMI-1 Monoclonal antibody specifically detects BMI-1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

BMI-1
LLBmi1-1
Immunocytochemistry/Immunofluorescence
P35226
B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51Polycomb group RING finger protein 4
Mouse
IgG2b Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
Purified
BMI1
Human Bmi1 protein [Swiss-Prot# P35226]
42 kDa
0.1 ml
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, Hematopoietic Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary
648
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only