Learn More
anti-BOB1, Clone: 1170CT1.1.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232301
Catalog No. NBP232301
Add to cart
Description
Description
BOB1 Monoclonal antibody specifically detects BOB1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
BOB1
1170CT1.1.1
Unconjugated
PBS with 0.09% Sodium Azide
Purified
POU2AF1
Purified His-tagged POU2AF1 protein was used to produced this monoclonal antibody.
27.436 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot
0.5 mg/mL
Western Blot 1:4000
Q16633
B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1
Mouse
IgG1 Kappa
Protein G purified
RUO
Primary
5450