Learn More
anti-BRCA1, Clone: 9F12, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP141214
Catalog No. NBP141214
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
BRCA1
9F12
Unconjugated
PBS (pH 7.2) with No Preservative
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1
Mouse
IgG1
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody will recognize human BRCA1.
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
1.0mg/mL
Western Blot 1-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation
Purified
BRCA1
BRCA1 protein fragment expressed in E. coli corresponding to amino acids 341-748.
220 kDa
100 ul
Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger
Primary
672
Human