Learn More
anti-BRCA2, Clone: 9D3, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP141187
Catalog No. NBP141187
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
BRCA2
9D3
Unconjugated
PBS (pH 7.2) with No Preservative
BRCA1/BRCA2-containing complex, subunit 2, BRCC2, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast cancer susceptibility protein BRCA2, breast cancer type 2 susceptibility protein, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1Fanconi anemia, complementation group D1, Fanconi anemia group D1 protein, GLM3, PNCA2
Mouse
IgG1
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody will recognize human BRCA2.
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
1.0mg/mL
Western Blot 1-5 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500
Purified
BRCA2
BRCA2 protein fragment containing amino acids 2336-2478 of human BRCA2 expressed in E. coli. BRCA2-P400.
390 kDa
100 ul
Breast Cancer, Cancer
Primary
675
Human