Learn More
anti-Bu-1a/b, FITC, Clone: 21-1A4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128320
Catalog No. NBP128320
Add to cart
Description
Description
Bu-1a Monoclonal antibody specifically detects Bu-1a in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
Specifications
Bu-1a/b
21-1A4
FITC
PBS with 0.05% Sodium Azide
Mouse
IgG1 Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Chicken
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Purified
Bursa cells from one-day-old H.B15 (Bu-1a/b) strain chickens
Protein A or G purified
RUO
Primary
Chicken Bu-1a/b alloantigen, Mr 70 kDa