Learn More
anti-Bu-1b, Clone: 5-11G2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128323
Catalog No. NBP128323
Add to cart
Description
Description
Bu-1b Monoclonal antibody specifically detects Bu-1b in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
Specifications
Bu-1b
5-11G2
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
B6.1, B6.2, Bu 1b, LOC396098
Bursa cells from two- to five-week-old CHA (Bu-1b) strain chickens
Protein A or G purified
RUO
Primary
Chicken Bu-1b alloantigen, Mr 70 kDa
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
0.5 mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunocytochemistry
Purified
Mouse
IgG1 Kappa
0.5 mg
Store at 4C. Do not freeze.
Monoclonal
Chicken