Learn More
Bu1 Mouse anti-Human, Chicken, Clone: AV20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128340
Catalog No. NBP128340
Add to cart
Description
Description
Bu1 Monoclonal antibody specifically detects Bu1 in Human, Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry.
Specifications
Specifications
Bu1
AV20
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
B6.1, B6.2, Bu 1a, Bu 1b, LOC396098
Chicken peripheral blood lymphocytes and splenocytes
Protein A or G purified
RUO
Primary
Chicken Bu-1 (chB6), Mr 70-75 kDa
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
0.5 mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunocytochemistry
Purified
Mouse
IgG1 Kappa
0.5 mg
Store at 4C. Do not freeze.
Monoclonal
Chicken, Human