Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-C-Peptide, Clone: HCP-B2, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP150685

Catalog No. NBP150685

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C-Peptide
HCP-B2
Unconjugated
PBS with 0.05% Sodium Azide
Affinity Purified
INS
Full length synthetic human C-Peptide
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500
P01308
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
IgG1
0.1 mg
Diabetes Research
Primary
3630
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only