Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse Calcitonin, Clone: 4901, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130051

Catalog No. NBP130051


Add to cart

Description

Description

Calcitonin Monoclonal antibody specifically detects Calcitonin in Human, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot (Negative), Immunocytochemistry.
Specifications

Specifications

Calcitonin
4901
Immunocytochemistry/Immunofluorescence 1:50-1:100, Western Blot (Negative), Immunocytochemistry 1:10 - 1:500
P01256
ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump
Mouse
IgG2a
0.1 ml
Cancer, Cellular Markers, Neuroscience
Primary
796
Human, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Purified
CALCA
Purified rat alpha-CGRP
Protein G purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Specific for Calcitonin Gene Related Peptide. Also recognizes rat beta-CGRP and human alpha-CGRP (10-fold less sensitivity compared to rat alpha-CGRP). Suggested positive controls are dorsal root ganglia, pancreas and gut.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only