Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cas9 Mouse anti-Mouse, Bacteria, DyLight 680, Clone: 7A9-3A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236440FR

Catalog No. NBP236440FR


Add to cart

Description

Description

CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in Mouse, Bacteria samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

CRISPR-Cas9
7A9-3A3
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Bacteria, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2]
Protein G purified
RUO
Primary
Cas9 protein from Streptococcus pyogene serotype M1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only