Learn More
anti-Caspase-14, Clone: 70A1426, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233245
Catalog No. NBP233245
Add to cart
Description
Description
Caspase-14 Monoclonal antibody specifically detects Caspase-14 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Caspase-14
70A1426
Western Blot 2-4 ug/ml
Ascites
CASP14
This antibody was raised against a synthetic peptide corresponding to amino acids 2-18 of mouse Caspase-14.
Unpurified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
In mouse spleen, a 30 kDa band is observed.
Western Blot
Unconjugated
Ascites with PBS - BSA free with No Preservative
apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE
Mouse
IgG1 Kappa
0.1 ml
Apoptosis, Cancer, Caspases
Primary
23581
Human, Mouse