Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cav1.2 Mouse anti-Human, Mouse, Rat, Hamster, Rabbit, Alexa Fluor 647, Clone: S57-46, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122439AF647
Description
Cav1.2 Monoclonal antibody specifically detects Cav1.2 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
Cav1.2 | |
S57-46 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Purified | |
CACNA1C | |
Fusion protein amino acids 1507-1733 (intracellular carboxyl terminus) of rabbit Cav1.2 | |
Protein G purified | |
RUO | |
Store at 4C in the dark. | |
Monoclonal | |
Detects approx 240kDa (varies with cell background due to glycosylation). |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
alpha-1 subunit, calcium channel, voltage-dependent, L type, alpha 1C subunit, cardic dihydropyridine-sensitive, alpha-1 subunit, L type, alpha 1 polypeptide, isoform 1, cardic muscle, L type, alpha-1 polypeptide, isoform 1, cardiac muscle, splice variant 10* | |
Mouse | |
IgG1 | |
0.1 ml | |
Phospho Specific | |
Primary | |
775 | |
Hamster, Human, Mouse, Rabbit, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only