Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse CCL2/MCP1, Clone: MNA1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142360

Catalog No. NBP142360


Add to cart

Description

Description

CCL2/MCP1 Monoclonal antibody specifically detects CCL2/MCP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay, Block/Neutralize.
Specifications

Specifications

CCL2/MCP1
MNA1
Unconjugated
PBS containing 0.1% BSA. with No Preservative
C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF
Mouse
IgG1
0.2 ml
Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
Primary
6347
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay, Blocking/Neutralizing 50-200 molar excess
Purified
CCL2
The antibody reacts with both human natural and recombinant MCP1 in ELISA.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The antibody reacts with both human natural and recombinant MCP1 in ELISA. MCP1 is a member of the CC subfamily of chemokines. MCP1 is a proinflammatory cytokine which acts both as an activation factor and as chemokine for monocytic cells. The antibody inhibits the biological activity of natural and recombinant human MCP1. The antigen specificity was assessed by ELISA. The antigen specificity was further assessed by immunoprecipitation and western blot. No crossreactivities with other cytokines have been detected.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only