Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse CCR2, Alexa Fluor 405, Clone: 2D8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP235333AF405

Catalog No. NBP235333W


Add to cart

Description

Description

CCR2 Monoclonal antibody specifically detects CCR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CCR2
2D8
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CCR2
A synthetic peptide made to an N-terminal portion of the human CCR2 protein (between residues 1-100) [Uniprot: P41597]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Immunogen's sequence similarity with other species: Mouse (57%), Rabbit (57%).
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
Mouse
IgG2b Lambda
0.1 ml
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary
729230
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only