Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR2 Antibody (3G7), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP235334V
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR2 | |
Monoclonal | |
DyLight 405 | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Immunogen's sequence similarity with other species: Mouse (57%), Rabbit (57%). | |
Store at 4C in the dark. | |
IgG2b λ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3G7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CCR2 | |
A synthetic peptide made to an N-terminal portion of the human CD63 protein (between residues 1-100) [UniProt P41597] | |
0.1 mL | |
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
729230 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction