Learn More
anti-CD1.1 antigen, DyLight 350, Clone: CB3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128362UV
Catalog No. NBP128362UV
Add to cart
Description
Description
CD1.1 antigen Monoclonal antibody specifically detects CD1.1 antigen in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD1.1 antigen
CB3
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CD1A
Lymphocytes from the bursa of Fabricius of outbred chickens
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Chicken CD1.1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a
Mouse
IgG1 Kappa
0.1 ml
Dendritic Cell Markers, Immunology
Primary
909
Chicken