Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1 Antibody (76-7-4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128221APC
Description
CD1 Monoclonal specifically detects CD1 in Porcine samples. It is validated for Flow Cytometry.Specifications
CD1 | |
Monoclonal | |
APC | |
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Porcine CD1 | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry | |
76-7-4 | |
Flow Cytometry | |
CD1A | |
Fresh dd miniature swine thymocytes | |
0.25 mL | |
Dendritic Cell Markers, Immunology | |
909 | |
Porcine | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction