Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD1, Biotin, Clone: 76-7-4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128223

Catalog No. NBP128223


Add to cart

Description

Description

CD1 Monoclonal antibody specifically detects CD1 in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications

Specifications

CD1
76-7-4
Biotin
PBS with 0.1% Sodium Azide
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a
Mouse
IgG2a Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Porcine CD1
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunocytochemistry
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Purified
CD1A
Fresh dd miniature swine thymocytes
Protein A or G purified
RUO
Primary
909
Porcine
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only