Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD103 Rat anti-Mouse, Brilliant Violet 421, Clone: M290, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 562771

Catalog No. BDB562771


Add to cart

Description

Description

The M290 antibody specificaly binds to CD103, the α chain of αIELβ7 integrin. CD103 has a unique and fairly restricted tissue distribution. It is expressed on almost all intestinal intraepithelial lymphocytes (IEL), on dendritic epidermal T cells (DEC), on subpopulations of peripheral T cells, and on distinct subsets of fetal, neonatal, and adult thymocytes. E-cadherin is the epithelial-cell ligand for αIELβ7 integrin. The ordered expression of αIEL during thymocyte development (which occurs under the influence of the thymic epithelium), the high level of expression of ααIELβ7-mediated adhesion in in vitro assays.

Specifications

Specifications

CD103
M290
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Itgae; Integrin alpha-E; αE; alpha-E1; ITAE; Integrin αIEL chain; aM290
Affinity Purified
Mouse Intestinal Epithelial Cells
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit