Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse CD105, PE-CF594, Clone: MJ7/18; BD

Manufacturer:  BD Biosciences 562762

Catalog No. BDB562762


Add to cart

Description

Description

Host Species: Rat
Clone: MJ7/18
Isotype: IgG2a, κ
Cross Reactivity Species: Ms
Immunogen: Mouse skin (inflamed)

Flow Cytometry

Specifications

Specifications

Mouse CD105
0.2mg/mL
Affinity Purified
Mouse skin (inflamed)
50μg
Primary
Murine
MJ7/18
PE-CF594
Rat
IgG2a
RUO
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit