Learn More
anti-Mouse CD117/c-kit, Biotin, Clone: kit2c75, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128329
Catalog No. NBP128329
Add to cart
Description
Description
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Chicken samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD117/c-kit
kit2c75
Biotin
PBS with 0.05% Sodium Azide
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Mouse
IgG2a Kappa
0.25 mg
Hematopoietic Stem Cell Markers, Mast Cell Markers, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
3815
Chicken
Flow Cytometry
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells
Purified
KIT
Sp/chkit4A4 cells (Sp2/0 cells transfected with chicken c-Kit cDNA)
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Chicken c-Kit