Learn More
anti-CD15/Lewis X, Clone: BRA4F1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142198
Catalog No. NBP142198
Add to cart
Description
Description
SSEA-1 Monoclonal antibody specifically detects SSEA-1 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
CD15/Lewis X
BRA4F1
Unconjugated
PBS with 0.05% sodium azide with No Preservative
CD 15, CD-15, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Mouse
IgM
0.1 mg
Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers
Primary
2526
Mouse
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
0.1 mg/mL
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
FUT4
The antibody reacts with the CD15 antigen.
IgM purified
RUO
Store at 4C. Do not freeze.
Monoclonal
This CD15 (BRA4F1) specifically detects CD15. CD15 contains the pentasaccharide lacto-N-fucopentatose III and is particularly expressed on granulocytes and mature neutrophils and on a wide variety of tumor cells including myeloid leukemia, breast, colorectal, and lung cancer cells. This CD15 (BRA4F1) was clustered at the IVth International Workshop on Leucocyte Differentiation Antigens. Source: A BALB/c mouse was immunized with K562 (an erythroid-myeloid leukemia cell line). Splenocytes were fused with mouse myeloma SP2.0-Ag14 cells.